missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorosalicylic acid, MedChemExpress

Product Code. 16449386
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16449386 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16449386 Supplier Medchem Express Supplier No. HYW016867
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 4-Chlorosalicylic acid is a pharmaceutical intermediate. Inhibits monophenolase and diphenolase activity with IC50s of 1.89 mM and 1.10 mM. Potent antimicrobial activity. Against E. coli with the MIC of 250 μg/mL and with the MBC of 500 μg/mL.

4-Chlorosalicylic acid is a pharmaceutical intermediate. Inhibits monophenolase and diphenolase activity with IC50s of 1.89 mM and 1.10 mM. Potent antimicrobial activity. Against E. coli with the MIC of 250 μg/mL and with the MBC of 500 μg/mL.

Specifications

Chemical Name or Material 4-Chlorosalicylic acid
CAS 5106-98-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C7H5ClO3
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (579.51 mM; Need ultrasonic) ∣H2O : 1 mg/mL (5.80 mM; ultrasonic and warming and heat to 80°C)
SMILES C1=CC(=CC(=C1C(O)=O)O)Cl
Molecular Weight (g/mol) 172.56
Formula Weight 172.56
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.