missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chlorosalicylic acid, 98%

Product Code. 11420717
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420717 100 g 100g
11410717 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420717 Supplier Alfa Aesar™ Supplier No. B22520.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5106-98-9
Molecular Formula C7H5ClO3
Molecular Weight (g/mol) 172.56
MDL Number MFCD00002449
InChI Key LWXFCZXRFBUOOR-UHFFFAOYSA-N
Synonym 4-chlorosalicylic acid, 4-chloro salicylic acid, benzoic acid, 4-chloro-2-hydroxy, 2-hydroxy-4-chlorobenzoic acid, 4-chloro-2-hydroxy benzoic acid, 4-chloro-2-hydroxy-benzoic acid, salicylic acid, 4-chloro, 4-chlorosalicylicacid, pubchem3845, 4-chloro-salicylic acid
PubChem CID 78782
IUPAC Name 4-chloro-2-hydroxybenzoic acid
SMILES OC(=O)C1=CC=C(Cl)C=C1O

Specifications

Melting Point ∼215°C (decomposition)
Quantity 100 g
Beilstein 2209125
Formula Weight 172.57
Percent Purity 98%
Chemical Name or Material 4-Chlorosalicylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.