missing translation for 'onlineSavingsMsg'
Learn More

4-Chlororesorcinol, 98%

Product Code. 11491067
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11491067 500 g 500g
11471067 25 g 25g
11481067 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491067 Supplier Thermo Scientific Alfa Aesar Supplier No. B24076.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-88-5 | C6H5ClO2 | 144.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 95-88-5
Molecular Formula C6H5ClO2
Molecular Weight (g/mol) 144.55
MDL Number MFCD00002273
InChI Key JQVAPEJNIZULEK-UHFFFAOYSA-N
Synonym 4-chlororesorcinol, 1,3-benzenediol, 4-chloro, p-chlororesorcinol, 4-chlororesorcin, resorcinol, 4-chloro, 6-chlororesorcinol, 4-chloro-1,3-dihydroxybenzene, 4-chloro-1,3-benzenediol, 2,4-dihydroxychlorobenzene, 1,3-dihydroxy-4-chlorobenzene
PubChem CID 1731
IUPAC Name 4-chlorobenzene-1,3-diol
SMILES OC1=CC=C(Cl)C(O)=C1

Specifikationer

Melting Point 108°C to 111°C
Boiling Point 146°C to 148°C (18 mmHg)
Quantity 500 g
Beilstein 2042864
Formula Weight 144.56
Percent Purity 98%
Chemical Name or Material 4-Chlororesorcinol

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.