missing translation for 'onlineSavingsMsg'
Learn More

4-Chlororesorcinol, 98%

Product Code. 11471067
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471067 25 g 25g
11481067 100 g 100g
11491067 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11471067 Supplier Thermo Scientific Alfa Aesar Supplier No. B24076.14
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 95-88-5 | C6H5ClO2 | 144.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-88-5
Molecular Formula C6H5ClO2
Molecular Weight (g/mol) 144.55
MDL Number MFCD00002273
InChI Key JQVAPEJNIZULEK-UHFFFAOYSA-N
Synonym 4-chlororesorcinol, 1,3-benzenediol, 4-chloro, p-chlororesorcinol, 4-chlororesorcin, resorcinol, 4-chloro, 6-chlororesorcinol, 4-chloro-1,3-dihydroxybenzene, 4-chloro-1,3-benzenediol, 2,4-dihydroxychlorobenzene, 1,3-dihydroxy-4-chlorobenzene
PubChem CID 1731
IUPAC Name 4-chlorobenzene-1,3-diol
SMILES OC1=CC=C(Cl)C(O)=C1

Specifications

Melting Point 108°C to 111°C
Boiling Point 146°C to 148°C (18 mmHg)
Quantity 25 g
Beilstein 2042864
Formula Weight 144.56
Percent Purity 98%
Chemical Name or Material 4-Chlororesorcinol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.