missing translation for 'onlineSavingsMsg'
Learn More

4-Chlororesorcinol, 98%

Product Code. 10316270 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10316270 100 g Glass bottle 100g
10489590 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10316270 Supplier Thermo Scientific Supplier No. 110061000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-88-5 | C6H5ClO2 | 144.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-88-5
Molecular Formula C6H5ClO2
Molecular Weight (g/mol) 144.55
MDL Number MFCD00002273
InChI Key JQVAPEJNIZULEK-UHFFFAOYSA-N
Synonym 4-chlororesorcinol, 1,3-benzenediol, 4-chloro, p-chlororesorcinol, 4-chlororesorcin, resorcinol, 4-chloro, 6-chlororesorcinol, 4-chloro-1,3-dihydroxybenzene, 4-chloro-1,3-benzenediol, 2,4-dihydroxychlorobenzene, 1,3-dihydroxy-4-chlorobenzene
PubChem CID 1731
IUPAC Name 4-chlorobenzene-1,3-diol
SMILES OC1=CC=C(Cl)C(O)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 106°C to 110°C
Color Beige
Boiling Point 147°C (18.0 mmHg)
Flash Point 148°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 g
Beilstein 06, II, 818
Solubility Information Solubility in water: 1000g/L (25°C)
Formula Weight 144.56
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Chlororesorcinol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chlororesorcinol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.