missing translation for 'onlineSavingsMsg'
Learn More

4-chloroquinoline, 99%

Product Code. 10441094 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10441094 1 g 1g
10567444 5 g 5g
11956491 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10441094 Supplier Thermo Scientific Acros Supplier No. 110050010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 611-35-8 | C9H6ClN | 163.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-35-8
Molecular Formula C9H6ClN
Molecular Weight (g/mol) 163.60
MDL Number MFCD00006773
InChI Key KNDOFJFSHZCKGT-UHFFFAOYSA-N
Synonym quinoline, 4-chloro, 4-chloro-quinoline, ccris 3979, 4-chioroquinoline, acmc-209mpi, 4-chloroquinoline, ksc354e5n, 4-chloroquinoline 1g, kndofjfshzckgt-uhfffaoysa
PubChem CID 69140
IUPAC Name 4-chloroquinoline
SMILES ClC1=C2C=CC=CC2=NC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 29.0°C to 32.0°C
Density 1.2500g/mL
Boiling Point 260.0°C to 261.0°C
Flash Point 110°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Beilstein 20,36
Specific Gravity 1.25
Formula Weight 163.61
Percent Purity 99%
Chemical Name or Material 4-chloroquinoline
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chloroquinoline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.