missing translation for 'onlineSavingsMsg'
Learn More

4-Chloroquinoline, 99%

Product Code. 15463248
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15463248 5 g 5g
15453248 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15463248 Supplier Thermo Scientific Alfa Aesar Supplier No. H56721.06
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 611-35-8 | C9H6ClN | 163.60 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Partly soluble in water.

Notes
Incompatible with oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-35-8
Molecular Formula C9H6ClN
Molecular Weight (g/mol) 163.60
MDL Number MFCD00006773
InChI Key KNDOFJFSHZCKGT-UHFFFAOYSA-N
Synonym quinoline, 4-chloro, 4-chloro-quinoline, ccris 3979, 4-chioroquinoline, acmc-209mpi, 4-chloroquinoline, ksc354e5n, 4-chloroquinoline 1g, kndofjfshzckgt-uhfffaoysa
PubChem CID 69140
IUPAC Name 4-chloroquinoline
SMILES ClC1=C2C=CC=CC2=NC=C1

Specifications

Melting Point 28°C to 31°C
Density 1.25
Boiling Point 260°C to 261°C
Flash Point 113°C (235°F)
Quantity 5 g
Beilstein 114721
Solubility Information Partly soluble in water.
Formula Weight 163.6
Percent Purity 99%
Chemical Name or Material 4-Chloroquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.