missing translation for 'onlineSavingsMsg'
Learn More

4-Chloropyrazole, 98%

Product Code. 15579503 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15579503 25 g 25g
15569503 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15579503 Supplier Thermo Scientific Acros Supplier No. 459920250
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15878-00-9 | C3H3ClN2 | 102.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15878-00-9
Molecular Formula C3H3ClN2
Molecular Weight (g/mol) 102.52
InChI Key BADSZRMNXWLUKO-UHFFFAOYSA-N
Synonym 4-chloropyrazole, 1h-pyrazole, 4-chloro, 4-chloro pyrazole, 1h-pyrazole, 4-chloro-9ci, pyrazole, 4-chloro, zlchem 159, pubchem14005, 5-23-04-00164 beilstein handbook reference
PubChem CID 27524
IUPAC Name 4-chloro-1H-pyrazole
SMILES C1=C(C=NN1)Cl

Specifications

Melting Point 71.0°C to 73.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Formula Weight 102.52
Percent Purity 98%
Chemical Name or Material 4-Chloropyrazole
compliance-icons
Product Identifier
  • 4-Chloropyrazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.