missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloropropiophenone, 98%, Thermo Scientific Chemicals

Catalog Number p-7081582
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
6285-05-8
MFCD00000626
This item is not returnable. View return policy
6285-05-8
MFCD00000626

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 33°C to 36°C
Boiling Point 125°C to 126°C (13 mmHg)
Flash Point >110°C (230°F)
Molecular Formula C7H12O
Quantity 25 g
Beilstein 1100638
Synonym 4'-chloropropiophenone, 1-4-chlorophenyl propan-1-one, p-chloropropiophenone, 4-chloropropiophenone, 1-propanone, 1-4-chlorophenyl, propiophenone, 4'-chloro, propiophenone, p-chloro, ethyl p-chlorophenyl ketone, usaf ek-5296, unii-26ay8wja1eAfficher moins
Solubility Information Insoluble in water
InChI Key PUNRPAWKFTXZIW-UHFFFAOYSA-N
SMILES CCC(CC)(C#C)O
IUPAC Name 3-ethylpent-1-yn-3-ol
Molecular Weight (g/mol) 112.17
PubChem CID 22698
Formula Weight 168.62
Percent Purity 98%
Chemical Name or Material 4'-Chloropropiophenone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'