missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenylacetyl chloride, 98%

Product Code. 11454194
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11454194

Brand: Thermo Scientific Alfa Aesar A12530.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25026-34-0 | C8H6Cl2O | 189.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25026-34-0
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.04
MDL Number MFCD00037111
InChI Key UMQUIRYNOVNYPA-UHFFFAOYSA-N
Synonym 4-chlorophenylacetyl chloride, p-chlorophenylacetyl chloride, 2-4-chlorophenyl acetyl chloride, 4-chlorobenzeneacetyl chloride, benzeneacetyl chloride, 4-chloro, 4-chlorophenylacetylchloride, acmc-1cch1, 4-chlorobenzeneacetylchloride, 4-chlorobenzenacetyl chloride, p-chlorophenyl-acetyl chloride
PubChem CID 90692
IUPAC Name 2-(4-chlorophenyl)acetyl chloride
SMILES ClC(=O)CC1=CC=C(Cl)C=C1

Specifications

Density 1.292
Boiling Point 112°C to 114°C (11 mmHg)
Quantity 10 g
Refractive Index 1.551
UN Number UN3265
Beilstein 637775
Formula Weight 189.04
Percent Purity 98%
Chemical Name or Material 4-Chlorophenylacetyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.