missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenylacetonitrile, 98+%

Product Code. 11488156
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11488156 100 g 100g
11498156 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11488156 Supplier Thermo Scientific Alfa Aesar Supplier No. A15295.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 140-53-4 | C8H6ClN | 151.593 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-53-4
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.593
MDL Number MFCD00001918
InChI Key IVYMIRMKXZAHRV-UHFFFAOYSA-N
Synonym 4-chlorobenzyl cyanide, 4-chlorophenyl acetonitrile, 4-chlorophenylacetonitrile, 2-4-chlorophenyl acetonitrile, p-chlorobenzyl cyanide, 4-chlorobenzeneacetonitrile, benzeneacetonitrile, 4-chloro, p-chlorophenyl acetonitrile, p-chlorophenylacetonitrile, 4-chlorobenzylcyanide
PubChem CID 241582
ChEBI CHEBI:17346
IUPAC Name 2-(4-chlorophenyl)acetonitrile
SMILES C1=CC(=CC=C1CC#N)Cl

Specifications

Melting Point 28°C to 31°C
Density 1.19
Boiling Point 141°C to 143°C (11 mmHg)
Flash Point 134°C (273°F)
Odor Pungent
Refractive Index 1.543
Quantity 100 g
UN Number UN3439
Beilstein 971171
Formula Weight 151.6
Percent Purity ≥98%
Chemical Name or Material 4-Chlorophenylacetonitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.