missing translation for 'onlineSavingsMsg'
Learn More

4-chlorophenylacetone, 98%

Product Code. 10351534 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10351534

Brand: Thermo Scientific Acros 350700250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5586-88-9 | C9H9ClO | 168.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5586-88-9
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00045214
InChI Key WEJRYKSUUFKMBC-UHFFFAOYSA-N
Synonym 4-chlorophenylacetone, 1-4-chlorophenyl acetone, 1-4-chlorophenyl propan-2-one, 1-p-chlorophenyl acetone, 1-4-chloro-phenyl-propan-2-one, p-chlorophenyl acetone, 2-propanone, 1-4-chlorophenyl, p-chlorophenylacetone, 4-chlorophenyl acetone
PubChem CID 79699
IUPAC Name 1-(4-chlorophenyl)propan-2-one
SMILES CC(=O)CC1=CC=C(Cl)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.151g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Refractive Index 1.5325 to 1.5345 (20°C, 589nm)
Quantity 25 g
Beilstein 07, IV, 691
Formula Weight 168.62
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-chlorophenylacetone
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.