missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenylacetic acid, 98%

Product Code. 11427187
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11427187 50 g 50g
11437187 250 g 250g
11417187 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11427187 Supplier Thermo Scientific Alfa Aesar Supplier No. A14974.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1878-66-6 | C8H7ClO2 | 170.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1878-66-6
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00004344
InChI Key CDPKJZJVTHSESZ-UHFFFAOYSA-N
Synonym 4-chlorophenylacetic acid, 2-4-chlorophenyl acetic acid, 4-chlorophenyl acetic acid, 4-chlorobenzeneacetic acid, p-chlorophenylacetic acid, benzeneacetic acid, 4-chloro, p-chlorophenyl acetic acid, 2-p-chlorophenyl acetic acid, acetic acid, p-chlorophenyl
PubChem CID 15880
ChEBI CHEBI:30749
IUPAC Name 2-(4-chlorophenyl)acetic acid
SMILES OC(=O)CC1=CC=C(Cl)C=C1

Specifications

Melting Point 103°C to 107°C
Quantity 50 g
Odor Odorless
Beilstein 1072816
Formula Weight 170.6
Percent Purity 98%
Chemical Name or Material 4-Chlorophenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.