missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenylacetic acid, 98%

Product Code. 10655982 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10655982 100 g Plastic bottle 100g
10038851 5 g Glass bottle 5g
10387860 500 g Plastic bottle 500g
10046043 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10655982 Supplier Thermo Scientific Acros Supplier No. 160721000
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1878-66-6 | C8H7ClO2 | 170.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1878-66-6
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00004344
InChI Key CDPKJZJVTHSESZ-UHFFFAOYSA-N
Synonym 4-chlorophenylacetic acid, 2-4-chlorophenyl acetic acid, 4-chlorophenyl acetic acid, 4-chlorobenzeneacetic acid, p-chlorophenylacetic acid, benzeneacetic acid, 4-chloro, p-chlorophenyl acetic acid, 2-p-chlorophenyl acetic acid, acetic acid, p-chlorophenyl
PubChem CID 15880
ChEBI CHEBI:30749
IUPAC Name 2-(4-chlorophenyl)acetic acid
SMILES OC(=O)CC1=CC=C(Cl)C=C1

Specifications

Melting Point 103°C to 107°C
CAS Min % 97.5
Color White to Cream
CAS Max % 100.0
Quantity 100 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Powder
Assay Percent Range 97.5% min. (HPLC)
Linear Formula ClC6H4CH2CO2H
Beilstein 09,448
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone,diethyl ether,solubility 5% in chcl3: clear solution
Formula Weight 170.6
Percent Purity 98%
Chemical Name or Material 4-Chlorophenylacetic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.