missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-chlorophenyl)methanol, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
50 g
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Linear Formula | ClC6H4CH2OH |
Quantity | 50 g |
Formula Weight | 142.58 |
Percent Purity | 97% |
Chemical Name or Material | (4-Chlorophenyl)methanol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'