missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-chlorophenyl)methanesulfonyl chloride, 90%, Thermo Scientific™
Click to view available options
Amber Glass Bottle; 1g
Specifications
CAS | 6966-45-6 |
CAS Min % | 90.0 |
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Molecular Formula | C7H6Cl2O2S |
MDL Number | MFCD01631927 |
Quantity | 1 g |
Synonym | 4-chlorophenyl methanesulfonyl chloride, 4-chlorobenzylsulfonyl chloride, 4-chloro-phenyl-methanesulfonyl chloride, 4-chloro-alpha-toluenesulfonyl chloride, chloro 4-chlorophenyl methyl sulfone, 4-chlorobenzenemethanesulfonyl chloride, 4-chlorophenyl methanesulphonyl chloride, 4-chlorophenyl methyl sulfonyl chloride, acmc-20apnh |
InChI Key | DBJRPJSDYFDWPV-UHFFFAOYSA-N |
SMILES | C1=CC(=CC=C1CS(=O)(=O)Cl)Cl |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'