missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenyl isocyanate, 98%

Product Code. 10215740 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10215740 25 g Glass bottle 25g
10154370 100 g Glass bottle 100g
10341144 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10215740 Supplier Thermo Scientific Acros Supplier No. 166560250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104-12-1 | C7H4ClNO | 153.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-12-1
Molecular Formula C7H4ClNO
Molecular Weight (g/mol) 153.57
MDL Number MFCD00002024
InChI Key ADAKRBAJFHTIEW-UHFFFAOYSA-N
Synonym 4-chlorophenyl isocyanate, p-chlorophenyl isocyanate, benzene, 1-chloro-4-isocyanato, pcpi, 4-chloroisocyanatobenzene, para-chlorophenyl isocyanate, p-chlorfenylisokyanat, 4-chlorophenylisocyanate, isocyanic acid, p-chlorophenyl ester, 1-chloro-4-isocyanato-benzene
PubChem CID 7693
IUPAC Name 1-chloro-4-isocyanatobenzene
SMILES C1=CC(=CC=C1N=C=O)Cl

Specifications

Melting Point 29.0°C to 31.0°C
CAS Min % 97.5
Color White to Yellow
CAS Max % 100.0
Density 1.2600g/mL
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Solid
Refractive Index 1.5618
Assay Percent Range 97.5% min. (GC)
Specific Gravity 1.26
Linear Formula ClC6H4NCO
Beilstein 12, 616
Boiling Point 203.0°C to 204.0°C
Solubility Information Solubility in water: decomposes in water
Formula Weight 153.57
Percent Purity 98%
Chemical Name or Material 4-Chlorophenyl isocyanate
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Chlorophenyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: 4-Chlorophenyl isocyanate

RUO – Research Use Only

Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.