missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenyl isocyanate, 98%

Product Code. 11499130
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499130 25 g 25g
11489130 5 g 5g
11409140 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499130 Supplier Thermo Scientific Alfa Aesar Supplier No. A14966.14
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 104-12-1 | C7H4ClNO | 153.565 g/mol

4-Chlorophenyl isocyanate is used in the preparation of isothiocyanates. Aryl isocyanates undergo [2+2] cycloaddition reactions with dihydrofuran to give bicyclic β-lactams in high yield.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorophenyl isocyanate is used in the preparation of isothiocyanates. Aryl isocyanates undergo [2+2] cycloaddition reactions with dihydrofuran to give bicyclic ß-lactams in high yield.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-12-1
Molecular Formula C7H4ClNO
Molecular Weight (g/mol) 153.565
MDL Number MFCD00002024
InChI Key ADAKRBAJFHTIEW-UHFFFAOYSA-N
Synonym 4-chlorophenyl isocyanate, p-chlorophenyl isocyanate, benzene, 1-chloro-4-isocyanato, pcpi, 4-chloroisocyanatobenzene, para-chlorophenyl isocyanate, p-chlorfenylisokyanat, 4-chlorophenylisocyanate, isocyanic acid, p-chlorophenyl ester, 1-chloro-4-isocyanato-benzene
PubChem CID 7693
IUPAC Name 1-chloro-4-isocyanatobenzene
SMILES C1=CC(=CC=C1N=C=O)Cl

Specifications

Melting Point 29°C to 31°C
Density 1.2
Quantity 25 g
Odor Pungent
Refractive Index 1.5618
UN Number UN2811
Beilstein 386235
Boiling Point 203°C to 204°C
Solubility Information Hydrolyzes in water.
Formula Weight 153.57
Percent Purity 98%
Chemical Name or Material 4-Chlorophenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.