missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenyl isocyanate, 98%

Product Code. 10341144 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10341144 500 g Glass bottle 500g
10215740 25 g Glass bottle 25g
10154370 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10341144 Supplier Thermo Scientific Acros Supplier No. 166565000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 104-12-1 | C7H4ClNO | 153.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-12-1
Molecular Formula C7H4ClNO
Molecular Weight (g/mol) 153.57
MDL Number MFCD00002024
InChI Key ADAKRBAJFHTIEW-UHFFFAOYSA-N
Synonym 4-chlorophenyl isocyanate, p-chlorophenyl isocyanate, benzene, 1-chloro-4-isocyanato, pcpi, 4-chloroisocyanatobenzene, para-chlorophenyl isocyanate, p-chlorfenylisokyanat, 4-chlorophenylisocyanate, isocyanic acid, p-chlorophenyl ester, 1-chloro-4-isocyanato-benzene
PubChem CID 7693
IUPAC Name 1-chloro-4-isocyanatobenzene
SMILES C1=CC(=CC=C1N=C=O)Cl

Specifications

Melting Point 29.0°C to 31.0°C
CAS Min % 97.5
Color White to Yellow
CAS Max % 100.0
Density 1.2600g/mL
Quantity 500 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Solid
Refractive Index 1.5618
Assay Percent Range 97.5% min. (GC)
Specific Gravity 1.26
Linear Formula ClC6H4NCO
Beilstein 12, 616
Boiling Point 203.0°C to 204.0°C
Solubility Information Solubility in water: decomposes in water
Formula Weight 153.57
Percent Purity 98%
Chemical Name or Material 4-Chlorophenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.