missing translation for 'onlineSavingsMsg'
Learn More

(4-Chlorophenyl)(4-piperidyl)methanone hydrochloride, 95%, Thermo Scientific™

Product Code. 10202642
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10202642 1 g Amber Glass Bottle 1g
10631483 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10202642 Supplier Thermo Scientific Maybridge Supplier No. CD10000DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 55695-51-7
Molecular Formula C12H15ClNO
Molecular Weight (g/mol) 224.71
MDL Number MFCD00053028
InChI Key IYGWDOXHCPQXKN-UHFFFAOYSA-O
Synonym 4-chlorophenyl piperidin-4-yl methanone hydrochloride, 4-4-chlorobenzoyl piperidine hydrochloride, 4-chlorophenyl 4-piperidyl methanone hydrochloride, 4-chlorophenyl 4-piperidinyl methanone hydrochloride, 4-p-chlorobenzoyl piperidine hydrochloride, 4-4-chlorophenyl carbonyl piperidine hydrochloride, methanone, 4-chlorophenyl-4-piperidinyl-, hydrochloride, 4-chlorophenyl 4-piperidinyl methanonehydrochloride, bestipharma 579-258, 4-4-chlorobenzoyl piperidine hcl
PubChem CID 2777114
IUPAC Name (4-chlorophenyl)-piperidin-4-ylmethanone;hydrochloride
SMILES ClC1=CC=C(C=C1)C(=O)C1CC[NH2+]CC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 260.16
Percent Purity 95%
Chemical Name or Material (4-Chlorophenyl)(4-piperidyl)methanone hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.