missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenoxyacetonitrile, 98%, Thermo Scientific Chemicals

Product Code. 11428346
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11428346 25 g 25g
11418346 5 g 5g
11438346 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428346 Supplier Thermo Scientific Alfa Aesar Supplier No. A15389.14
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3598-13-8 | Molecular Formula: C8H6ClNO | Molecular Weight: 167.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3598-13-8
Molecular Formula C8H6ClNO
Molecular Weight (g/mol) 167.59
MDL Number MFCD00031535
InChI Key YUGDKEWUYZXXRU-UHFFFAOYSA-N
Synonym 2-4-chlorophenoxy acetonitrile, 4-chlorophenoxyacetonitrile, 4-chlorophenoxy acetonitrile, 4-chloro-phenoxy-acetonitrile, p-chlorophenoxyacetonitrile, 1-chloro-4-cyanomethoxy benzene, acetonitrile, 4-chlorophenoxy, 4-chloro-o-cyanomethylphenol, 2-4-chlorophenoxy ethanenitrile, acmc-20aopp
PubChem CID 306444
IUPAC Name 2-(4-chlorophenoxy)acetonitrile
SMILES ClC1=CC=C(OCC#N)C=C1

Specifications

Melting Point 45°C to 49°C
Quantity 25 g
UN Number UN3439
Beilstein 2442163
Boiling Point 149°C to 150°C (15 mmHg)
Formula Weight 167.6
Percent Purity 98%
Chemical Name or Material 4-Chlorophenoxyacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.