missing translation for 'onlineSavingsMsg'
Learn More

4-chlorophenoxyacetonitrile, 98%, ACROS Organics™

Product Code. 10832631 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10832631 25 g 25g
10833971 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10832631 Supplier Thermo Scientific Acros Supplier No. 437600250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 3598-13-8
Molecular Formula C8H6ClNO
Molecular Weight (g/mol) 167.59
MDL Number MFCD00031535
InChI Key YUGDKEWUYZXXRU-UHFFFAOYSA-N
Synonym 2-4-chlorophenoxy acetonitrile, 4-chlorophenoxyacetonitrile, 4-chlorophenoxy acetonitrile, 4-chloro-phenoxy-acetonitrile, p-chlorophenoxyacetonitrile, 1-chloro-4-cyanomethoxy benzene, acetonitrile, 4-chlorophenoxy, 4-chloro-o-cyanomethylphenol, 2-4-chlorophenoxy ethanenitrile, acmc-20aopp
PubChem CID 306444
IUPAC Name 2-(4-chlorophenoxy)acetonitrile
SMILES ClC1=CC=C(OCC#N)C=C1

Specifica

Melting Point 45°C to 49°C
CAS Min % 97.5
Color Tan-Yellow
CAS Max % 100.0
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Powder
Assay Percent Range 98%
Linear Formula ClC6H4OCH2CN
Formula Weight 167.6
Percent Purity ≥97.5%
Chemical Name or Material 4-chlorophenoxyacetonitrile
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.