missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenoxyacetic acid, 98%

Product Code. 10627142 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10627142 500 g Plastic bottle 500g
10499080 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10627142 Supplier Thermo Scientific Acros Supplier No. 166965000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 122-88-3 | C8H7ClO3 | 186.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-88-3
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.59
MDL Number MFCD00004305
InChI Key SODPIMGUZLOIPE-UHFFFAOYSA-N
Synonym 4-chlorophenoxyacetic acid, 2-4-chlorophenoxy acetic acid, 4-chlorophenoxy acetic acid, 4-cpa, sure-set, tomatotone, tomato fix, acetic acid, 4-chlorophenoxy, p-chlorophenoxyacetic acid, tomato hold
PubChem CID 26229
ChEBI CHEBI:1808
IUPAC Name 2-(4-chlorophenoxy)acetic acid
SMILES C1=CC(=CC=C1OCC(=O)O)Cl

Specifications

Melting Point >156.0°C
CAS Min % 97.5
Color White to Beige
CAS Max % 100.0
Quantity 500 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Powder
Assay Percent Range 98%
Linear Formula ClC6H4OCH2CO2H
Beilstein 06, 187
Solubility Information Solubility in water: sparingly soluble. Other solubilities: soluble in most common organic solvents
Formula Weight 186.59
Percent Purity 98%
Chemical Name or Material 4-Chlorophenoxyacetic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chlorophenoxyacetic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.