missing translation for 'onlineSavingsMsg'
Få mere at vide

(4-Chlorophenoxy)acetic Acid, Mikromol™

Artikelnummer. 30050984
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
25 mg
100 mg
Pakningsstørrelse:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
30050984 25 mg 25mg
17943396 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30050984 Leverandør Mikromol Leverandørnr. MM0044.020025
 Restricted Item
Estimated Shipment: 04-08-2026
Læg i indkøbskurv
Læg i indkøbskurv
Denne vare kan ikke returneres. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(4-Chlorophenoxy)acetic Acid
TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material 4-Chlorophenoxyacetic acid
CAS 122-88-3
Analyte or Component Names 4-Chlorophenoxyacetic acid
Shipping Condition Room Temperature
Molecular Formula C8 H7 Cl O3
Certifications/Compliance ISO 9001
Synonym (4-Chlorophenoxy)acetic Acid
InChI Formula InChI=1S/C8H7ClO3/c9-6-1-3-7(4-2-6)12-5-8(10)11/h1-4H,5H2,(H,10,11)
SMILES OC(=O)COc1ccc(Cl)cc1
IUPAC Name 2-(4-chlorophenoxy)acetic acid
Molecular Weight (g/mol) 186.59
Quantity 25 mg
Formula Weight 186.0084 g/mol
Vis mere Vis mindre
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.