missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenol, TRC

Product Code. 30136191
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136191 25 g 25g
17939629 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136191 Supplier Toronto Research Chemicals Supplier No. TRCC37504025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-48-9
Molecular Formula C6 H5 Cl O
Molecular Weight (g/mol) 128.56
Synonym Phenol, p-chloro- (8CI), 4-Chlorophenol, 4-CP, 4-Hydroxychlorobenzene, Applied 3-78, NSC 2877, p-Chlorophenic acid, p-Chlorophenol
IUPAC Name 4-chlorophenol
SMILES Oc1ccc(Cl)cc1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C6H5ClO/c7-5-1-3-6(8)4-2-5/h1-4,8H
Formula Weight 128.0029
Chemical Name or Material 4-Chlorophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.