missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenol, 99%, Thermo Scientific Chemicals

Product Code. 11457467
Click to view available options
Quantity:
100 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as main raw material of compounding for medicine, dye, plastic and other industries. 4-Chlorophenol was used in visible-light-induced degradation of 4-chlorophenol in aqueous suspension of pure TiO2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as main raw material of compounding for medicine, dye, plastic and other industries. 4-Chlorophenol was used in visible-light-induced degradation of 4-chlorophenol in aqueous suspension of pure TiO2.

Solubility
Soluble in water (27g/L).

Notes
Air sensitive. Store in a cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-48-9
Molecular Formula C6H5ClO
Molecular Weight (g/mol) 128.555
MDL Number MFCD00002318
InChI Key WXNZTHHGJRFXKQ-UHFFFAOYSA-N
Synonym p-chlorophenol, parachlorophenol, phenol, 4-chloro, 4-hydroxychlorobenzene, phenol, p-chloro, applied 3-78, 4-monochlorophenol, p-chlorophenic acid, p-chlorfenol, 4-chloro-1-hydroxybenzene
PubChem CID 4684
ChEBI CHEBI:28078
IUPAC Name 4-chlorophenol
SMILES C1=CC(=CC=C1O)Cl

Specifications

Melting Point 39°C to 43°C
Density 1.306
Boiling Point 217°C to 220°C
Flash Point 115°C (239°F)
Odor Phenol-like
Quantity 1000 g
UN Number UN2020
Beilstein 507004
Sensitivity Air Sensitive
Merck Index 14,2154
Solubility Information Soluble in water (27g/L).
Formula Weight 128.56
Percent Purity 99%
Chemical Name or Material 4-Chlorophenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'