missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)styrene (~0.1% TBC stabilizer), TRC

Product Code. 30352568
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
25g
Product Code. Quantity unitSize
30352568 25 g 25g
30365102 10 g -
2 options
This item is not returnable. View return policy

Product Code. 30352568

Brand: Toronto Research Chemicals TRCC58798325G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1592-20-7
Molecular Formula C9H9Cl
Molecular Weight (g/mol) 152.62
IUPAC Name 1-(chloromethyl)-4-ethenylbenzene
SMILES ClCc1ccc(C=C)cc1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C9H9Cl/c1-2-8-3-5-9(7-10)6-4-8/h2-6H,1,7H2
Formula Weight 152.0393
Chemical Name or Material 4-(Chloromethyl)styrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.