missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)benzyl alcohol, 97%, Thermo Scientific Chemicals

Product Code. 11994651 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11994651 1 g 1g
11904661 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11994651 Supplier Thermo Scientific Acros Supplier No. 438920010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16473-35-1
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.61
InChI Key OGALXJIOJZXBBP-UHFFFAOYSA-N
Synonym 4-chloromethyl benzyl alcohol, 4-chloromethyl phenyl methanol, 4-chloromethyl-phenyl-methanol, 4-chloromethylbenzyl alcohol, benzenemethanol, 4-chloromethyl, 4-chloromethyl benzylalcohol, acmc-20a8hi, p-hydroxymethylbenzyl chloride, 4-hydroxymethylbenzyl chloride
PubChem CID 5107660
IUPAC Name [4-(chloromethyl)phenyl]methanol
SMILES C1=CC(=CC=C1CO)CCl

Specifications

Melting Point 58.0°C to 60.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2C6H4CH2OH
Quantity 1 g
Formula Weight 156.61
Percent Purity 97%
Chemical Name or Material 4-(Chloromethyl)benzyl alcohol
compliance-icons
Product Identifier
  • 4-(Chloromethyl)benzyl alcohol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.