missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)benzoic Acid, TRC

Product Code. 30350517
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30350517 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350517 Supplier Toronto Research Chemicals Supplier No. TRCC3691035G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1642-81-5
Molecular Formula C8H7ClO2
Quantity 5 g
Synonym 4-(Chloromethyl)benzenecarboxylic Acid, NSC 123935, p-(Chloromethyl)benzoic Acid, alpha-Chloro-p-toluic Acid
InChI Formula InChI=1S/C8H7ClO2/c9-5-6-1-3-7(4-2-6)8(10)11/h1-4H,5H2,(H,10,11)
SMILES OC(=O)c1ccc(CCl)cc1
IUPAC Name 4-(chloromethyl)benzoic acid
Molecular Weight (g/mol) 170.59
Formula Weight 170.0135
Chemical Name or Material 4-(Chloromethyl)benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.