missing translation for 'onlineSavingsMsg'
Learn More

4-Chloromethyl-5-methyl-2-phenyl-oxazole, TRC

Product Code. 30358768
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358768 100 mg 100mg
30354799 50 mg 50mg
30348803 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30358768 Supplier Toronto Research Chemicals Supplier No. TRCC382883100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103788-61-0
Molecular Formula C11H10NOCl
Molecular Weight (g/mol) 207.65
Synonym 5-Methyl-2-phenyl-4-(chloromethyl)oxazole, [5-Methyl-2-phenyloxazol-4-yl]methyl Chloride, 4-(Chloromethyl)-5-methyl-2-phenyloxazole, 4-(Chloromethyl)-5-methyl-2-phenyl-1,3-oxazole, 4-Chloromethyl-2-phenyl-5-methyloxazole
IUPAC Name 4-(chloromethyl)-5-methyl-2-phenyl-1,3-oxazole
SMILES Cc1oc(nc1CCl)c2ccccc2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H10ClNO/c1-8-10(7-12)13-11(14-8)9-5-3-2-4-6-9/h2-6H,7H2,1H3
Formula Weight 207.0451
Chemical Name or Material 4-Chloromethyl-5-methyl-2-phenyl-oxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.