missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)-3-(2,6-dichlorophenyl)-5-methylisoxazole, 97%, Thermo Scientific™

Product Code. 10529592
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10529592 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10529592 Supplier Thermo Scientific Maybridge Supplier No. SPB07543CB
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 303225-22-1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H8Cl3NO
MDL Number MFCD00833265
Quantity 250 mg
Synonym 4-chloromethyl-3-2,6-dichlorophenyl-5-methylisoxazole, 4-chloromethyl-3-2,6-dichlorophenyl-5-methyl-1,2-oxazole, pubchem8711, 3-2,6-dichlorophenyl-4-chloromethyl-5-methylisoxazole, isoxazole,4-chloromethyl-3-2,6-dichlorophenyl-5-methyl, 2-4-chloromethyl-5-methyl-1,2-oxazol-3-yl-1,3-dichlorobenzene, 3-2,6-bis chloranyl phenyl-4-chloromethyl-5-methyl-1,2-oxazole
InChI Key PPUGTWAPVDDBEW-UHFFFAOYSA-N
SMILES CC1=C(C(=NO1)C2=C(C=CC=C2Cl)Cl)CCl
IUPAC Name 4-(chloromethyl)-3-(2,6-dichlorophenyl)-5-methyl-1,2-oxazole
Molecular Weight (g/mol) 276.541
PubChem CID 2735759
Formula Weight 276.55
Percent Purity 97%
Chemical Name or Material 4-(Chloromethyl)-3-(2, 6-dichlorophenyl)-5-methylisoxazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.