missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Chloromethyl)-2-methyl-1,3-thiazole hydrochloride , Tech., Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Specifications
Specifications
| Quantity | 5 g |
| Formula Weight | 184.09 |
| Percent Purity | Tech. |
| Grade | Technical |
| Chemical Name or Material | 4-(Chloromethyl)-2-methyl-1, 3-thiazole hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction