missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)-2-(4-chlorophenyl)-1,3-thiazole, 97%, Thermo Scientific™

Product Code. 10455122
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10455122 1 g Amber Glass Bottle 1g
10782483 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10455122 Supplier Thermo Scientific Maybridge Supplier No. CD03309DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 17969-22-1
Molecular Formula C10H7Cl2NS
Molecular Weight (g/mol) 244.133
MDL Number MFCD00047057
InChI Key UEJQTBKTWJQBRN-UHFFFAOYSA-N
Synonym 4-chloromethyl-2-4-chlorophenyl-1,3-thiazole, thiazole, 4-chloromethyl-2-4-chlorophenyl, 4-chloromethyl-2-4-chlorophenyl thiazole, thiazole,4-chloromethyl-2-4-chlorophenyl, 4-chloromethyl-2-4-chloro-phenyl-thiazole, thiazole, 4-chloromethyl-2-p-chlorophenyl, 4-chloromethyl-2-p-chlorophenylthiazole, 4-chloromethyl-2-4-chlorphenyl thiazole, 2-p-chlorophenyl-4-chloromethylthiazole
PubChem CID 610297
IUPAC Name 4-(chloromethyl)-2-(4-chlorophenyl)-1,3-thiazole
SMILES C1=CC(=CC=C1C2=NC(=CS2)CCl)Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 244.14
Percent Purity 95%
Chemical Name or Material 4-(Chloromethyl)-2-(4-chlorophenyl)-1, 3-thiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.