missing translation for 'onlineSavingsMsg'
Learn More

4-Chloroiodobenzene, TRC

Product Code. 30350617
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350617 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350617 Supplier Toronto Research Chemicals Supplier No. TRCC36829525G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Chloroiodobenzene
CAS 637-87-6
Molecular Formula C6H4ClI
Quantity 25 g
Synonym 1-Chloro-4-iodobenzene, 4-Chloro-1-iodobenzene, 4-Chlorophenyl Iodide, 4-Iodo-1-chlorobenzene, 4-Iodochlorobenzene, NSC 32862, p-Chloroiodobenzene, p-Chlorophenyl Iodide, p-Iodochlorobenzene, p-Iodophenyl Chloride
InChI Formula InChI=1S/C6H4ClI/c7-5-1-3-6(8)4-2-5/h1-4H
SMILES Clc1ccc(I)cc1
IUPAC Name 1-chloro-4-iodobenzene
Molecular Weight (g/mol) 238.45
Formula Weight 237.9046
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.