missing translation for 'onlineSavingsMsg'
Learn More

4-(Chlorocarbonyl)benzoic Acid Methyl Ester, TRC

Product Code. 30350068
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350068 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350068 Supplier Toronto Research Chemicals Supplier No. TRCC36605010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(Chlorocarbonyl)benzoic Acid Methyl Ester
CAS 7377-26-6
Molecular Formula C9H7ClO3
Quantity 10 g
Synonym p-(Chloroformyl)benzoic Acid Methyl Ester, 1-(Chloroformyl)-4-(methoxycarbonyl)benzene, 4-(Carbomethoxy)benzoyl Chloride, 4-(Methoxycarbonyl)benzoyl chloride, Methyl 4-(chlorocarbonyl)benzoate, Methyl 4-carboxybenzoyl Chloride, Methyl p-(chlorocarbonyl)benzoate, Methyl terephthalic Chloride, Methyl Terephthalyl Chloride, Monomethyl Terephthalate Acid Chloride, Terephthalic Acid Monomethyl Ester chloride, Terephthaloyl Chloride Methyl Ester, p-(Carbomethoxy)benzoyl Chloride
InChI Formula InChI=1S/C9H7ClO3/c1-13-9(12)7-4-2-6(3-5-7)8(10)11/h2-5H,1H3
SMILES COC(=O)c1ccc(cc1)C(=O)Cl
IUPAC Name methyl 4-carbonochloridoylbenzoate
Molecular Weight (g/mol) 198.6
Formula Weight 198.0084
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.