missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobutyrophenone, 95%, Thermo Scientific™

Product Code. 11362496
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362496 5 g 5g
11372496 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362496 Supplier Thermo Scientific Alfa Aesar Supplier No. L06383.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 939-52-6
Molecular Formula C10H11ClO
Molecular Weight (g/mol) 182.65
MDL Number MFCD00001005
InChI Key GHEFQKHLHFXSBR-UHFFFAOYSA-N
Synonym 4-chlorobutyrophenone, gamma-chlorobutyrophenone, 4-chloro-1-oxo-1-phenylbutane, .gamma.-chlorobutyrophenone, 1-butanone,4-chloro-1-phenyl, chlorpropylphenylketon, 4-chlorobutyrylbenzene, 4-chlorobutyro-phenone, 3-benzoylpropyl chloride, .omega.-chlorobutyrophenone
PubChem CID 253533
IUPAC Name 4-chloro-1-phenylbutan-1-one
SMILES ClCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 19°C to 20°C
Density 1.137
Boiling Point 138°C to 140°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.544
Quantity 5 g
Beilstein 878974
Formula Weight 182.65
Percent Purity 95%
Chemical Name or Material 4-Chlorobutyrophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.