missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobutyronitrile, TRC

Product Code. 30355909
Click to view available options
Quantity:
50 g
Unit Size:
50g
This item is not returnable. View return policy

Product Code. 30355909

Brand: Toronto Research Chemicals TRCC36504150G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Chlorobutyronitrile
CAS 628-20-6
Molecular Formula C4 H6 Cl N
Quantity 50 g
InChI Formula InChI=1S/C4H6ClN/c5-3-1-2-4-6/h1-3H2
SMILES ClCCCC#N
IUPAC Name 4-chlorobutanenitrile
Molecular Weight (g/mol) 103.55
Formula Weight 103.0189
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.