missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobutyl Acetate, TRC

Product Code. 30350063
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350063 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350063 Supplier Toronto Research Chemicals Supplier No. TRCC3648055G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Chlorobutyl Acetate
CAS 6962-92-1
Molecular Formula C6H11ClO2
Quantity 5 g
Synonym 4-Chloro-1-butanol Acetate, 1-Acetoxy-4-chlorobutane, 1-Chloro-4-acetoxybutane, 4-Chlorobutyl Acetate, Acetic Acid 4-Chlorobutyl Ester, NSC 53492, NSC 54074, delta-Chlorobutyl Acetate
InChI Formula InChI=1S/C6H11ClO2/c1-6(8)9-5-3-2-4-7/h2-5H2,1H3
SMILES CC(=O)OCCCCCl
IUPAC Name 4-chlorobutyl acetate
Molecular Weight (g/mol) 150.6
Formula Weight 150.0448
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.