missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobutyl acetate, 98%

Product Code. 11465157
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11465157 25 g 25g
11475157 100 g 100g
11485157 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11465157 Supplier Thermo Scientific Alfa Aesar Supplier No. B22582.14
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 6962-92-1 | C6H11ClO2 | 150.602 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6962-92-1
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150.602
MDL Number MFCD00001013
InChI Key PYLDCZJUHYVOAF-UHFFFAOYSA-N
Synonym 1-acetoxy-4-chlorobutane, 4-chloro-n-butyl acetate, 1-chloro-4-acetoxybutane, 1-butanol, 4-chloro-, acetate, 4-chlorobutylacetate, 4-chloro-1-butanol acetate, 4-chlorbut-1-ylacetat, delta-chlorobutyl acetate, 1-butanol, 4-chloro-, 1-acetate, 4-chlorbut-1-ylacetat german
PubChem CID 23399
IUPAC Name 4-chlorobutyl acetate
SMILES CC(=O)OCCCCCl

Specifications

Density 1.076
Boiling Point 81°C to 83°C (11 mmHg)
Flash Point 64°C (147°F)
Refractive Index 1.435
Quantity 25 g
Beilstein 1749681
Formula Weight 150.61
Percent Purity 98%
Chemical Name or Material 4-Chlorobutyl acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.