missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobutanoic acid methyl ester, TRC

Product Code. 30364910
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30364910 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 30364910

Brand: Toronto Research Chemicals TRCC29284510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Chlorobutanoic acid methyl ester
CAS 3153-37-5
Molecular Formula C5H9ClO2
Quantity 10 g
Synonym 4-Chlorobutyric acid methyl ester, Methyl 4-chlorobutanoate,Methyl 4-chlorobutyrate,
InChI Formula InChI=1S/C5H9ClO2/c1-8-5(7)3-2-4-6/h2-4H2,1H3
SMILES COC(=O)CCCCl
IUPAC Name methyl 4-chlorobutanoate
Molecular Weight (g/mol) 136.58
Formula Weight 136.0291
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.