missing translation for 'onlineSavingsMsg'
Learn More

4'-Chlorobiphenyl-4-sulfonyl chloride, 97%

Product Code. p-4813105 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 15299646

Brand: Thermo Scientific Acros 450330010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20443-74-7 | C12H8Cl2O2S | 287.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20443-74-7
Molecular Formula C12H8Cl2O2S
Molecular Weight (g/mol) 287.17
InChI Key NWYUSJMIHFIMTA-UHFFFAOYSA-N
Synonym 4'-chlorobiphenyl-4-sulfonyl chloride, 4-4-chlorophenyl benzenesulfonyl chloride, 4'-chloro-biphenyl-4-sulfonyl chloride, 4'-chloro-1,1'-biphenyl-4-sulfonyl chloride, 4'-chloro-4-biphenylylsulphonyl chloride, 4'-chloro 1,1'-biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 4'-chloro, 4'-chlorobiphenyl4-sulfonyl chloride, 4-chlorobiphenyl-4'-sulfonyl chloride, 4'-chloro biphenyl-4-sulfonyl chloride
PubChem CID 2794745
IUPAC Name 4-(4-chlorophenyl)benzenesulfonyl chloride
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)Cl)S(=O)(=O)Cl

Specifications

Melting Point 109.0°C to 110.0°C
Assay Percent Range 96% min. as diethylsulfonamide (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 287.17
Percent Purity 97%
Chemical Name or Material 4'-Chlorobiphenyl-4-sulfonyl chloride
compliance-icons
Product Identifier
  • 4'-Chlorobiphenyl-4-sulfonyl chloride, 97%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.