missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzylamine, 97+%

Product Code. 11421748
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11421748 100 g 100g
11344749 5 g 5g
11411748 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11421748 Supplier Thermo Scientific Alfa Aesar Supplier No. A13984.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 104-86-9 | C7H8ClN | 141.598 g/mol

4-Chlorobenzylamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzylamine is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-86-9
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.598
MDL Number MFCD00008121
InChI Key YMVFJGSXZNNUDW-UHFFFAOYSA-N
Synonym 4-chlorobenzylamine, 4-chlorophenyl methanamine, p-chlorobenzylamine, benzenemethanamine, 4-chloro, 1-4-chlorophenyl methanamine, 4-chloro-benzylamine, benzylamine, p-chloro, 4-chlorophenyl methylamine, para-chlorobenzylamine, chembl13218
PubChem CID 66036
IUPAC Name (4-chlorophenyl)methanamine
SMILES C1=CC(=CC=C1CN)Cl

Specifications

Density 1.16
Boiling Point 229°C
Flash Point 90°C (194°F)
Odor Amine-like
Refractive Index 1.559
Quantity 100 g
UN Number UN2735
Beilstein 507949
Sensitivity Air Sensitive
Solubility Information Not miscible in water.
Formula Weight 141.6
Percent Purity ≥97%
Chemical Name or Material 4-Chlorobenzylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.