missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzyl isothiocyanate, 97%

Product Code. 11337506
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11337506

Brand: Thermo Scientific Alfa Aesar L09579.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3694-45-9 | C8H6ClNS | 183.653 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3694-45-9
Molecular Formula C8H6ClNS
Molecular Weight (g/mol) 183.653
MDL Number MFCD00041116
InChI Key DEHXIHUIYSXZNH-UHFFFAOYSA-N
Synonym 4-chlorobenzyl isothiocyanate, 1-chloro-4-isothiocyanatomethyl benzene, p-chlorobenzyl isothiocyanate, benzene, 1-chloro-4-isothiocyanatomethyl, isothiocyanic acid, p-chlorobenzyl ester, 4-chlorobenzylisothiocyanate, benzene, 1-chloro-4-isothiocyanatomethyl-9ci, acmc-1cng5, wln: scn1r dg, p-chlorobenzylisothiocyanate
PubChem CID 19408
IUPAC Name 1-chloro-4-(isothiocyanatomethyl)benzene
SMILES C1=CC(=CC=C1CN=C=S)Cl

Specifications

Density 1.27
Boiling Point 169°C to 170°C (25 mmHg)
Quantity 5 g
Odor Irritating
Refractive Index 1.6145
UN Number UN2922
Beilstein 2085917
Formula Weight 183.66
Percent Purity 97%
Chemical Name or Material 4-Chlorobenzyl isothiocyanate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.