missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzyl cyanide, 98+%

Product Code. 10000890 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
250g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10000890 500 g Glass bottle 500g
10355740 5 g Glass bottle 5g
10080870 100 g Glass bottle 100g
10102760 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10000890 Supplier Thermo Scientific Acros Supplier No. 108815000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 140-53-4 | C8H6ClN | 151.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-53-4
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.6
MDL Number MFCD00001918
InChI Key IVYMIRMKXZAHRV-UHFFFAOYSA-N
Synonym 4-chlorobenzyl cyanide, 4-chlorophenyl acetonitrile, 4-chlorophenylacetonitrile, 2-4-chlorophenyl acetonitrile, p-chlorobenzyl cyanide, 4-chlorobenzeneacetonitrile, benzeneacetonitrile, 4-chloro, p-chlorophenyl acetonitrile, p-chlorophenylacetonitrile, 4-chlorobenzylcyanide
PubChem CID 241582
ChEBI CHEBI:17346
IUPAC Name 2-(4-chlorophenyl)acetonitrile
SMILES C1=CC(=CC=C1CC#N)Cl

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 29°C to 30°C
Color Colorless to Yellow
Boiling Point 265°C to 267°C
Flash Point 133°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4CH2CN
Refractive Index 1.5415 to 1.5435
Quantity 500 g
Beilstein 09, 448
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in acetone and alcohol
Formula Weight 151.6
Percent Purity 98+%
Physical Form Liquid After Melting
Chemical Name or Material 4-Chlorobenzyl cyanide, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chlorobenzyl cyanide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.