missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorobenzyl chloride, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
250 g
1000 g
5000 g
Description
4-Chlorobenzyl chloride was the test compound for determining rat hepatocellular thiols by HPLC. As a reagent used in derivatization of, for example, a calix[4]arene and a tryptamine. It is widely used as an intermediate for manufacturing organic compounds like benzyl alcohol and benzyl cyanide. Also used in fuel as a gum inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Chlorobenzyl chloride was the test compound for determining rat hepatocellular thiols by HPLC. As a reagent used in derivatization of, for example, a calix[4]arene and a tryptamine. It is widely used as an intermediate for manufacturing organic compounds like benzyl alcohol and benzyl cyanide. Also used in fuel as a gum inhibitor.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
4-Chlorobenzyl chloride was the test compound for determining rat hepatocellular thiols by HPLC. As a reagent used in derivatization of, for example, a calix[4]arene and a tryptamine. It is widely used as an intermediate for manufacturing organic compounds like benzyl alcohol and benzyl cyanide. Also used in fuel as a gum inhibitor.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
Specifications
Melting Point | 26°C to 30°C |
Density | 1.275 |
Boiling Point | 220°C to 221°C |
Flash Point | 107°C (225°F) |
Odor | Pungent |
Refractive Index | 1.5575 |
Quantity | 1000 g |
UN Number | UN3427 |
Beilstein | 471558 |
Solubility Information | Insoluble in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'