missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzyl bromide, 98+%

Product Code. 11449897
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449897 5 g 5g
11459897 25 g 25g
11469897 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449897 Supplier Thermo Scientific Alfa Aesar Supplier No. B22858.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 622-95-7 | C7H6BrCl | 205.48 g/mol

4-Chlorobenzyl bromide is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzyl bromide is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, bases and active metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-95-7
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.48
MDL Number MFCD00040714
InChI Key KQNBRMUBPRGXSL-UHFFFAOYSA-N
Synonym 4-chlorobenzyl bromide, 1-bromomethyl-4-chlorobenzene, p-chlorobenzyl bromide, 4-chlorobenzylbromide, benzene, 1-bromomethyl-4-chloro, alpha-bromo-4-chlorotoluene, 4cbb, p-chlorobenzylbromide, 4-chloro-benzylbromide
PubChem CID 69329
IUPAC Name 1-(bromomethyl)-4-chlorobenzene
SMILES ClC1=CC=C(CBr)C=C1

Specifications

Melting Point 46°C to 51°C
Boiling Point 107°C to 108°C (8 mmHg)
Odor Pungent
Quantity 5 g
UN Number UN3261
Beilstein 606497
Solubility Information Insoluble in water.
Formula Weight 205.49
Percent Purity ≥98%
Chemical Name or Material 4-Chlorobenzyl bromide
compliance-icons
Product Identifier
  • 4-Chlorobenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 1-(Bromomethyl)-4-chlorobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.