missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzoylacetonitrile, 98%

Product Code. 11479503
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479503 100 g 100g
11459503 5 g 5g
11469503 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479503 Supplier Thermo Scientific Alfa Aesar Supplier No. A12364.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4640-66-8 | C9H6ClNO | 179.603 g/mol

Pharmaceutical Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pharmaceutical Intermediates.

Solubility
Soluble in Dichloromethane, Ether, Ethyl Acetate and Methanol

Notes
Store at -20°C. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4640-66-8
Molecular Formula C9H6ClNO
Molecular Weight (g/mol) 179.603
MDL Number MFCD00051625
InChI Key JYOUFPNYTOFCSJ-UHFFFAOYSA-N
Synonym 4-chlorobenzoylacetonitrile, 3-4-chlorophenyl-3-oxopropanenitrile, 4-chlorophenacylcyanide, p-chlorophenacyl cyanide, 4-chlorobenzoyl acetonitrile, 3-4-chloro-phenyl-3-oxo-propionitrile, 4-chlorobenzoylacetonitirle, zlchem 1256, pubchem12045, acmc-1akag
PubChem CID 138308
IUPAC Name 3-(4-chlorophenyl)-3-oxopropanenitrile
SMILES C1=CC(=CC=C1C(=O)CC#N)Cl

Specifications

Melting Point 128°C to 132°C
Quantity 100 g
Beilstein 743368
Solubility Information Soluble in Dichloromethane,Ether,Ethyl Acetate and Methanol
Formula Weight 179.61
Percent Purity 98%
Chemical Name or Material 4-Chlorobenzoylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.