missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzoyl Chloride, TRC

Product Code. 30362966
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30362966 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30362966 Supplier Toronto Research Chemicals Supplier No. TRCC36444025G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Chlorobenzoyl Chloride
CAS 122-01-0
Molecular Formula C7H4Cl2O
Quantity 25 g
Synonym 4-Chloro-benzoyl Chloride, p-Chloro-benzoyl Chloride, 4-Chlorobenzoic Acid Chloride, p-Chlorobenzoyl Chloride, para-Chlorobenzoyl Chloride
InChI Formula InChI=1S/C7H4Cl2O/c8-6-3-1-5(2-4-6)7(9)10/h1-4H
SMILES ClC(=O)c1ccc(Cl)cc1
IUPAC Name 4-chlorobenzoyl chloride
Molecular Weight (g/mol) 175.01
Formula Weight 173.9639
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.