missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzotrichloride, 98+%

Product Code. 11425427
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11425427

Brand: Thermo Scientific Alfa Aesar B23446.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5216-25-1 | C7H4Cl4 | 229.909 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5216-25-1
Molecular Formula C7H4Cl4
Molecular Weight (g/mol) 229.909
MDL Number MFCD00000790
InChI Key LVZPKYYPPLUECL-UHFFFAOYSA-N
Synonym 4-chlorobenzotrichloride, 1-chloro-4-trichloromethyl benzene, p-chlorobenzotrichloride, benzene, 1-chloro-4-trichloromethyl, p-chlorophenyltrichloromethane, p-trichloromethylchlorobenzene, alpha,alpha,alpha,4-tetrachlorotoluene, unii-1up9gju33s, ccris 8902, p,alpha,alpha,alpha-tetrachlorotoluene
PubChem CID 21277
IUPAC Name 1-chloro-4-(trichloromethyl)benzene
SMILES C1=CC(=CC=C1C(Cl)(Cl)Cl)Cl

Specifications

Melting Point 5°C to 6°C
Density 1.495
Boiling Point 245°C
Quantity 25 g
Refractive Index 1.573
UN Number UN1760
Beilstein 1866549
Formula Weight 229.92
Percent Purity ≥98%
Chemical Name or Material 4-Chlorobenzotrichloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.