missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorobenzonitrile, 99%, Thermo Scientific Chemicals
18.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Melting Point | 91°C to 94°C |
CAS Min % | 98.5 |
Color | White to Yellow |
CAS Max % | 100.0 |
Density | 108g/mL |
Boiling Point | 223°C |
Quantity | 25 g |
Infrared Spectrum | Authentic |
Packaging | Plastic bottle |
Physical Form | Flakes or Chunks |
Mostrar más |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'